Anti-ADAP2 Goat Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA305819EA 3810 DKK
ORIGTA305819
Anti-ADAP2 Goat Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: ADAP2 (ArfGAP with dual PH domains 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR