Anti-TNKS Goat Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA305803EA 3810 DKK
ORIGTA305803
Anti-TNKS Goat Antibody
Antistoffer
Recommended Dilutions: Western Blot: 05-15?g/ml

Type: Primary
Antigen: TNKS (tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR