Anti-DLG4 Goat antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA305790EA 3810 DKK
ORIGTA305790
Anti-DLG4 Goat antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: DLG4 (discs, large homolog 4 (Drosophila))
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR