Anti-CPT1B Goat Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA305764EA 3810 DKK
ORIGTA305764
Anti-CPT1B Goat Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 3-5?g/ml

Type: Primary
Antigen: CPT1B (carnitine palmitoyltransferase 1B (muscle))
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR