Anti-PRMT5 Goat Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA305754EA 3810 DKK
ORIGTA305754
Anti-PRMT5 Goat Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: PRMT5 (protein arginine methyltransferase 5)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR