Anti-RGS13 Goat Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA305720EA 3830 DKK
ORIGTA305720
Anti-RGS13 Goat Antibody
Antistoffer
Recommended Dilutions: Western Blot: 05-15?g/ml

Type: Primary
Antigen: RGS13 (regulator of G-protein signaling 13)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR