Anti-RNF34 Goat Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA305687EA 3810 DKK
ORIGTA305687
Anti-RNF34 Goat Antibody
Antistoffer
Recommended Dilutions: Immunochemistry: 4-8?g/ml

Type: Primary
Antigen: RNF34 (ring finger protein 34, E3 ubiquitin protein ligase)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR