Anti-GGCX Goat antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA305624EA 3830 DKK
ORIGTA305624
Anti-GGCX Goat antibody
Antistoffer
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: GGCX (gamma-glutamyl carboxylase)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR