Anti-NPM1 Rabbit monoclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA303620EA 3550 DKK
ORIGTA303620
Anti-NPM1 Rabbit monoclonal antibody
Recommended Dilutions: Western Blot: 1:200000; Immunochemistry: 1:100-250; Flow Cytometry: 1:40; Immunoprecipitation: 1:60

Type: Primary
Antigen: NPM1 (nucleophosmin (nucleolar phosphoprotein B23, numatrin))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR