Anti-DACH1 Goat polyclonal antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA303348EA 3830 DKK
ORIGTA303348
Anti-DACH1 Goat polyclonal antibody
Antistoffer
Recommended Dilutions: ELISA: 116000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: DACH1 (dachshund family transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR