Anti-ACTL7B Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA303311EA 3830 DKK
ORIGTA303311
Anti-ACTL7B Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: ELISA: 1:128000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: ACTL7B (actin-like 7B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR