Anti-PDE11A Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA303261EA 3830 DKK
ORIGTA303261
Anti-PDE11A Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: ELISA: 1:2000; Western Blot: 05-15µg/ml

Type: Primary
Antigen: PDE11A (phosphodiesterase 11A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR