Anti-KCNJ11 Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA303230EA 3810 DKK
ORIGTA303230
Anti-KCNJ11 Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: ELISA: 1:64000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: KCNJ11 (potassium inwardly-rectifying channel, subfamily J, member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR