Anti-ADRB1 Goat Polyclonal Antibody
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGTA303089EA 3390 DKK
ORIGTA303089
Anti-ADRB1 Goat Polyclonal Antibody
Recommended Dilutions: ELISA: 1:128000; Western Blot: 03-1µg/ml
Type: Primary
Antigen: ADRB1 (adrenoceptor beta 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human