Anti-CD14 Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA303082EA 3810 DKK
ORIGTA303082
Anti-CD14 Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: ELISA: 1:128000 Western Blot: 1-3µg/ml

Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR