Anti-RICTOR Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA303018EA 3810 DKK
ORIGTA303018
Anti-RICTOR Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: ELISA: 1:4000 Western Blot: 2-3µg/ml

Type: Primary
Antigen: RICTOR (RPTOR independent companion of MTOR, complex 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR