Anti-TOM1L2 Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA302974EA 3810 DKK
ORIGTA302974
Anti-TOM1L2 Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.1-1?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: TOM1L2 (target of myb1-like 2 (chicken))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR