Anti-RXRA Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA302967EA 3830 DKK
ORIGTA302967
Anti-RXRA Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: ELISA: 1:16000; Western Blot: 05-3µg/ml

Type: Primary
Antigen: RXRA (retinoid X receptor, alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR