Anti-PARD6A Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA302913EA 3810 DKK
ORIGTA302913
Anti-PARD6A Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: ELISA: 1:8000 Western Blot: 10µg/ml; Immunochemistry: 5-10µg/ml

Type: Primary
Antigen: PARD6A (par-6 family cell polarity regulator alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR