Anti-OSBPL1A Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA302903EA 3810 DKK
ORIGTA302903
Anti-OSBPL1A Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: ELISA: 1:16000; Western Blot: 05-2µg/ml

Type: Primary
Antigen: OSBPL1A (oxysterol binding protein-like 1A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR