Anti-KLF16 Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA302801EA 3810 DKK
ORIGTA302801
Anti-KLF16 Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 2-4µg/ml

Type: Primary
Antigen: KLF16 (Kruppel-like factor 16)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR