Anti-RGS1 Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA302787EA 3810 DKK
ORIGTA302787
Anti-RGS1 Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 05-2µg/ml

Type: Primary
Antigen: RGS1 (regulator of G-protein signaling 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR