Anti-TBL1X / TBL1XR1 Goat Polyclonal Antibody
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGTA302762EA 4090 DKK
ORIGTA302762
Anti-TBL1X / TBL1XR1 Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 3-5µg/ml
Type: Primary
Antigen: TBL1X / TBL1XR1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human