Anti-ELMO2 Goat polyclonal antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA302760EA 3810 DKK
ORIGTA302760
Anti-ELMO2 Goat polyclonal antibody
Antistoffer
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: ELMO2 (engulfment and cell motility 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR