Anti-DOK3 Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA302734EA 3830 DKK
ORIGTA302734
Anti-DOK3 Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: DOK3 (docking protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR