Anti-CSK Goat polyclonal antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA302657EA 3830 DKK
ORIGTA302657
Anti-CSK Goat polyclonal antibody
Antistoffer
Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.5-2µg/ml; Immunochemistry: 0.1-0.3?g/ml

Type: Primary
Antigen: CSK (c-src tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR