Anti-MTM1 Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA302542EA 3810 DKK
ORIGTA302542
Anti-MTM1 Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: ELISA: 1:16000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: MTM1 (myotubularin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR