Anti-HSD11B1 Goat Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA302508EA 3810 DKK
ORIGTA302508
Anti-HSD11B1 Goat Polyclonal Antibody
Antistoffer
Recommended Dilutions: ELISA: 1:4000; Western Blot: 03-2µg/ml

Type: Primary
Antigen: HSD11B1 (hydroxysteroid (11-beta) dehydrogenase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR