Anti-EHD2 Goat polyclonal antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA302472EA 4110 DKK
ORIGTA302472
Anti-EHD2 Goat polyclonal antibody
Antistoffer
Recommended Dilutions: ELISA: 1:32000; Western Blot: 0003-001µg/ml

Type: Primary
Antigen: EHD2 (EH-domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR