Anti-CKB Goat polyclonal antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA302453EA 3810 DKK
ORIGTA302453
Anti-CKB Goat polyclonal antibody
Antistoffer
Recommended Dilutions: ELISA: 1:2000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: CKB (creatine kinase, brain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR