Anti-FANCC Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA301760EA 4340 DKK
ORIGTA301760
Anti-FANCC Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: FANCC (Fanconi anemia, complementation group C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR