Anti-YWHAG Mouse Monoclonal Antibody [clone: KC21]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA301660EA 4590 DKK
ORIGTA301660
Anti-YWHAG Mouse Monoclonal Antibody [clone: KC21]
Antistoffer
Recommended Dilutions: Western Blot: 1:3000

Type: Primary
Antigen: YWHAG (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR