Anti-LOX Rabbit Polyclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA301449EA 4780 DKK
ORIGTA301449
Anti-LOX Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: LOX (lysyl oxidase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR