Anti-ARNT Mouse Monoclonal Antibody [clone: H1beta234]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA301443EA 5200 DKK
ORIGTA301443
Anti-ARNT Mouse Monoclonal Antibody [clone: H1beta234]
Antistoffer
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: ARNT (aryl hydrocarbon receptor nuclear translocator)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR