Anti-BIRC5 Mouse Monoclonal Antibody [clone: 60.11]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGTA301428EA 4910 DKK
ORIGTA301428
Anti-BIRC5 Mouse Monoclonal Antibody [clone: 60.11]
Antistoffer
Recommended Dilutions: Western Blot: 2 ?g/ml
Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat