Anti-TTR Rabbit Monoclonal Antibody [clone: EP2929Y]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA301271EA 5590 DKK
ORIGTA301271
Anti-TTR Rabbit Monoclonal Antibody [clone: EP2929Y]
Antistoffer
Recommended Dilutions: Western Blot: 1:2500-1:5000; Immunoprecipitation: 1:60; Immunochemistry: 1:10-1:500; Flow Cytometry: 1:50

Type: Primary
Antigen: TTR (transthyretin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR