Anti-MTOR Rabbit Monoclonal Antibody [clone: Y391]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA300537EA 5760 DKK
ORIGTA300537
Anti-MTOR Rabbit Monoclonal Antibody [clone: Y391]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000-1:2000; Immunochemistry/Immunofluorescence: 1:50-1:100; Flow Cytometry: 1:50-1:80; Immunoprecipitation: 1:50

Type: Primary
Antigen: MTOR (mechanistic target of rapamycin (serine/threonine kinase))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR