Anti-TERT Rabbit monoclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA300468EA 0 DKK
ORIGTA300468
Anti-TERT Rabbit monoclonal antibody
Type: Primary
Antigen: TERT (telomerase reverse transcriptase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR