Anti-tYFP Mouse Monoclonal Antibody

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA150027EA 3460 DKK
ORIGTA150027
Anti-tYFP Mouse Monoclonal Antibody
Antistoffer
Type: Tag
Antigen: tYFP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR