Anti-Myc Tag None Monoclonal Antibody (Biotin)

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGTA150015EA 4060 DKK
ORIGTA150015
Anti-Myc Tag None Monoclonal Antibody (Biotin)
Antistoffer
Type: Tag
Antigen: Myc Tag (Myc tag)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: None
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR