Anti-ZBTB33 Mouse Monoclonal Antibody

Leverandør: Abgent
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABGEAT4570AEA 2620 DKK
ABGEAT4570A
Anti-ZBTB33 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: ZBTB33 (zinc finger and BTB domain containing 33)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR