Anti-UGT2B10 Mouse Monoclonal Antibody
Leverandør: Abgent
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABGEAT4460AEA 2620 DKK
ABGEAT4460A
Anti-UGT2B10 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: UGT2B10 (UDP glucuronosyltransferase 2 family, polypeptide B10)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human