Anti-UBE2L3 Mouse Monoclonal Antibody

Leverandør: Abgent
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABGEAT4439AEA 2620 DKK
ABGEAT4439A
Anti-UBE2L3 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: UBE2L3 (ubiquitin-conjugating enzyme E2L 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR