Anti-TSC22D4 Mouse Monoclonal Antibody

Leverandør: Abgent
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABGEAT4377AEA 2620 DKK
ABGEAT4377A
Anti-TSC22D4 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: TSC22D4 (TSC22 domain family, member 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR