Anti-TK1 Mouse Monoclonal Antibody
Leverandør: Abgent
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABGEAT4249AEA 2620 DKK
ABGEAT4249A
Anti-TK1 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: TK1 (thymidine kinase 1, soluble)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human