Anti-STK32C Mouse Monoclonal Antibody

Leverandør: Abgent
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABGEAT4081AEA 2620 DKK
ABGEAT4081A
Anti-STK32C Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: STK32C (serine/threonine kinase 32C)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR