Anti-SOX3 Mouse Monoclonal Antibody
Leverandør: Abgent
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABGEAT3997AEA 2620 DKK
ABGEAT3997A
Anti-SOX3 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: SOX3 (SRY (sex determining region Y)-box 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human