Anti-S100A1 Mouse Monoclonal Antibody
Leverandør: Abgent
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABGEAT3750AEA 2620 DKK
ABGEAT3750A
Anti-S100A1 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: S100A1 (S100 calcium binding protein A1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human