Anti-PPIE Mouse Monoclonal Antibody
Leverandør: Abgent
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABGEAT3401AEA 2620 DKK
ABGEAT3401A
Anti-PPIE Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: PPIE (peptidylprolyl isomerase E (cyclophilin E))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human