Anti-NXT1 Mouse Monoclonal Antibody
Leverandør: Abgent
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABGEAT3141AEA 2620 DKK
ABGEAT3141A
Anti-NXT1 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: NXT1 (NTF2-like export factor 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human